siRNA Details
Gene Symbol: C12orf40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135051.1 | 9 | 1441 | |||
GenBank mRNA | AK097445.1 | - | 9 | 1390 | ||
EU668334.1 | - | 9 | 1390 | |||
GQ472216.1 | - | 9 | 1390 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C12orf40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135051.1 | 9 | 1441 | |||
GenBank mRNA | AK097445.1 | - | 9 | 1390 | ||
EU668334.1 | - | 9 | 1390 | |||
GQ472216.1 | - | 9 | 1390 |