siRNA Details
| Gene Symbol: C12orf40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135051.1 | 14, 15 | 2097 | |||
| GenBank mRNA | AK097445.1 | - | 14, 15 | 2046 | ||
| EU668334.1 | - | 14, 15 | 2046 | |||
| GQ472216.1 | - | 14, 15 | 2046 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C12orf40| Gene Symbol: C12orf40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135051.1 | 14, 15 | 2097 | |||
| GenBank mRNA | AK097445.1 | - | 14, 15 | 2046 | ||
| EU668334.1 | - | 14, 15 | 2046 | |||
| GQ472216.1 | - | 14, 15 | 2046 | |||