siRNA Details
| Gene Symbol: LOC100129938 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK127489.1 | - | 1 | 1448 | ||
| Gene Symbol: LRRC4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011527520.2 | XP_011525822.1 | 1 | 1448 | ||

LOC100129938
Genomic Map