siRNA Details
Gene Symbol: CYSLTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282186.1 | NP_001269115.1 | 2 | 2585 | ||
NM_001282187.1 | NP_001269116.1 | 4 | 2755 | |||
NM_001282188.1 | NP_001269117.1 | 4 | 2732 | |||
NM_006639.3 | NP_006630.1 | 3 | 2672 | |||
GenBank mRNA | DQ131803.1 | - | 1 | 1229 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
