siRNA Details
| Gene Symbol: ALMS1-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046762.1 | 2 | 1096 | |||
| GenBank mRNA | BC041487.1 | - | 2 | 1103 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ALMS1-IT1| Gene Symbol: ALMS1-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046762.1 | 2 | 1096 | |||
| GenBank mRNA | BC041487.1 | - | 2 | 1103 | ||