siRNA Details
| Gene Symbol: LOC100505502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK094743.1 | - | 1 | 117 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA198427.1 | - | 1 | 117 | Chr. 10 - 31363643 - 31399755 [-] on Build GRCh38 | |

LOC100505502
Genomic Map