siRNA Details
| Gene Symbol: LOC100505502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011519798.2 | XP_011518100.1 | 1 | 1188 | ||
| GenBank mRNA | AK094743.1 | - | 2 | 1287 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100505502| Gene Symbol: LOC100505502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011519798.2 | XP_011518100.1 | 1 | 1188 | ||
| GenBank mRNA | AK094743.1 | - | 2 | 1287 | ||