siRNA Details
| Gene Symbol: IRS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005262220.2 | XP_005262277.1 | 2 | 4773 | ||
| XM_011531061.1 | XP_011529363.1 | 3 | 4799 | |||
| GenBank mRNA | AL832136.1 | - | 1 | 3188 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

IRS4| Gene Symbol: IRS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005262220.2 | XP_005262277.1 | 2 | 4773 | ||
| XM_011531061.1 | XP_011529363.1 | 3 | 4799 | |||
| GenBank mRNA | AL832136.1 | - | 1 | 3188 | ||