siRNA Details
| Gene Symbol: ZNF546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297763.1 | NP_001284692.1 | 7 | 7595 | ||
| NM_178544.4 | NP_848639.2 | 7 | 7673 | |||
| XM_011526898.2 | XP_011525200.1 | 6 | 8740 | |||
| XM_011526899.2 | XP_011525201.1 | 7 | 7585 | |||
| GenBank mRNA | AF087985.1 | - | 1 | 303 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU176936.1 | - | 1 | 519 | Chr. 19 - 40020169 - 40021011 [+] on Build GRCh38 | |

ZNF546
Genomic Map