siRNA Details
| Gene Symbol: GGTA1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AF378121.1 | - | LIT2093 alpha1,3GTP1 |
8 | 1755 | |
| AF378123.1 | - | LIT2095 alpha1,3GTP3 |
8 | 1804 | ||

GGTA1P
Genomic Map