siRNA Details
| Gene Symbol: ABCB9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011538098.2 | XP_011536400.1 | 1 | 1336 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AF461897.1 | - | 1 | 383 | Chr. 12 - 122951241 - 122952311 [-] on Build GRCh38 | |

ABCB9
Genomic Map