siRNA Details
| Gene Symbol: LINC01441 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110064.1 | 3 | 1732 | |||
| GenBank mRNA | AK056050.1 | - | 3 | 1733 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000433766.1 | - | 3 | 1732 | Chr. 20 - 55420336 - 55427197 [-] on Build GRCh38 | |

LINC01441
Genomic Map