siRNA Details
| Gene Symbol: LINC01441 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110064.1 | 1 | 42 | |||
| GenBank mRNA | AK056050.1 | - | 1 | 42 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA684966.1 | - | 1 | 42 | Chr. 20 - 55422091 - 55427197 [-] on Build GRCh38 | |

LINC01441
Genomic Map