siRNA Details
| Gene Symbol: LINC01441 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110064.1 | 3 | 1777 | |||
| GenBank mRNA | AK056050.1 | - | 3 | 1778 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01441| Gene Symbol: LINC01441 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110064.1 | 3 | 1777 | |||
| GenBank mRNA | AK056050.1 | - | 3 | 1778 | ||