siRNA Details
| Gene Symbol: COA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005249807.4 | XP_005249864.1 | 7 | 6572 | ||
| XM_006715752.2 | XP_006715815.1 | 8 | 6540 | |||
| XM_017012414.1 | XP_016867903.1 | 7 | 6499 | |||
| XM_017012415.1 | XP_016867904.1 | 8 | 6601 | |||
| XM_017012417.1 | XP_016867906.1 | 8 | 5003 | |||
| XM_017012418.1 | XP_016867907.1 | 7 | 4873 | |||
| XM_017012421.1 | XP_016867910.1 | 8 | 6723 | |||
| XM_017012422.1 | XP_016867911.1 | 8 | 6612 | |||
| XM_017012424.1 | XP_016867913.1 | 7 | 6515 | |||
| XM_017012430.1 | XP_016867919.1 | 8 | 4975 | |||
| GenBank mRNA | BX537586.1 | - | 1 | 14 | ||

COA1
Genomic Map