siRNA Details
| Gene Symbol: COA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005249807.4 | XP_005249864.1 | 7 | 8762 | ||
| XM_006715752.2 | XP_006715815.1 | 8 | 8730 | |||
| XM_017012414.1 | XP_016867903.1 | 7 | 8689 | |||
| XM_017012415.1 | XP_016867904.1 | 8 | 8791 | |||
| XM_017012417.1 | XP_016867906.1 | 8 | 7193 | |||
| XM_017012418.1 | XP_016867907.1 | 7 | 7063 | |||
| XM_017012421.1 | XP_016867910.1 | 8 | 8913 | |||
| XM_017012422.1 | XP_016867911.1 | 8 | 8802 | |||
| XM_017012424.1 | XP_016867913.1 | 7 | 8705 | |||
| XM_017012430.1 | XP_016867919.1 | 8 | 7165 | |||
| GenBank mRNA | BX537586.1 | - | 1 | 2226 | ||

COA1
Genomic Map