siRNA Details
| Gene Symbol: WWTR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017006121.1 | XP_016861610.1 | 2, 3 | 214 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

WWTR1| Gene Symbol: WWTR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017006121.1 | XP_016861610.1 | 2, 3 | 214 | ||