siRNA Details
| Gene Symbol: SLCO4A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017027827.1 | XP_016883316.1 | 12 | 3283 | ||
| GenBank mRNA | AK095985.1 | - | 1 | 1108 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLCO4A1| Gene Symbol: SLCO4A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017027827.1 | XP_016883316.1 | 12 | 3283 | ||
| GenBank mRNA | AK095985.1 | - | 1 | 1108 | ||