siRNA Details
| Gene Symbol: CLUL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011525651.2 | XP_011523953.1 | 1 | 280 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK128498.1 | - | 1112 | Chr. - - [-] on Build | ||

CLUL1
Genomic Map