siRNA Details
| Gene Symbol: CASP16P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK096989.1 | - | 2 | 186 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA922641.1 | - | 2 | 186 | Chr. 16 - 3144219 - 3145347 [+] on Build GRCh38 | |

CASP16P
Genomic Map