siRNA Details
| Gene Symbol: LOC142937 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC008131.2 | - | 1 | 220 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | HY207780.1 | - | 1 | 26 | Chr. 10 - 6171966 - 6172414 [+] on Build GRCh38 | |

LOC142937
Genomic Map