siRNA Details
| Gene Symbol: LINC01165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK097584.1 | - | 1 | 710 | ||
| Gene Symbol: LOC107984282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017017028.1 | XP_016872517.1 | 2 | 2995 | ||

LINC01165
Genomic Map