siRNA Details
Gene Symbol: LINC01165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK097584.1 | - | 1 | 1669 |
Gene Symbol: LOC107984282 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017017028.1 | XP_016872517.1 | 2 | 3954 |
