siRNA Details
| Gene Symbol: FLJ40536 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK097855.1 | - | 1 | 1903 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FLJ40536| Gene Symbol: FLJ40536 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK097855.1 | - | 1 | 1903 | ||