siRNA Details
| Gene Symbol: FER | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308028.1 | NP_001294957.1 | 18 | 9499 | ||
| NM_001308031.1 | NP_001294960.1 | 11 | 8449 | |||
| NM_005246.3 | NP_005237.2 | 20 | 9885 | |||
| XM_011543276.2 | XP_011541578.1 | 12 | 8701 | |||
| GenBank mRNA | AK021888.1 | - | 1 | 1318 | ||
| AK092320.1 | - | 1 | 2253 | |||
| AK123847.1 | - | 1 | 3114 | |||

FER
Genomic Map