siRNA Details
| Gene Symbol: C15orf59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039614.2 | NP_001034703.1 | 2 | 5406 | ||
| NM_001303254.1 | NP_001290183.1 | 3 | 5289 | |||
| XM_005254369.4 | XP_005254426.1 | 3 | 6689 | |||
| GenBank mRNA | AK124841.1 | - | 1 | 2789 | ||
| AL360257.1 | - | 1 | 1539 | |||

C15orf59
Genomic Map