siRNA Details
| Gene Symbol: C15orf59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039614.2 | NP_001034703.1 | 2 | 5204 | ||
| NM_001303254.1 | NP_001290183.1 | 3 | 5087 | |||
| XM_005254369.4 | XP_005254426.1 | 3 | 6487 | |||
| GenBank mRNA | AL360257.1 | - | 1 | 1337 | ||

C15orf59
Genomic Map