siRNA Details
| Gene Symbol: PLCL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK023546.1 | - | 5 | 2406 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK125854.1 | - | 1 | 2573 | Chr. 3 - 17078964 - 17082796 [+] on Build GRCh38 | |

PLCL2
Genomic Map