siRNA Details
| Gene Symbol: LOC79160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125359.1 | 1 | 575 | |||
| GenBank mRNA | BC002831.2 | - | 1 | 575 | 3636069 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC79160| Gene Symbol: LOC79160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125359.1 | 1 | 575 | |||
| GenBank mRNA | BC002831.2 | - | 1 | 575 | 3636069 | |