siRNA Details
| Gene Symbol: STARD4-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040093.1 | 6 | 2452 | |||
| GenBank mRNA | CR749489.1 | - | 6 | 2454 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

STARD4-AS1| Gene Symbol: STARD4-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040093.1 | 6 | 2452 | |||
| GenBank mRNA | CR749489.1 | - | 6 | 2454 | ||