siRNA Details
| Gene Symbol: TUBGCP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011537464.2 | XP_011535766.1 | 1 | 373 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK125472.1 | - | 1 | 348 | Chr. 13 - 112602828 - 112606417 [-] on Build GRCh38 | |

TUBGCP3
Genomic Map