siRNA Details
| Gene Symbol: LINC01023 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK126449.1 | - | 2 | 1129 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX117545.1 | - | 1 | 104 | Chr. 5 - 108726365 - 108727124 [-] on Build GRCh38 | |

LINC01023
Genomic Map