siRNA Details
| Gene Symbol: SLCO5A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005251313.2 | XP_005251370.1 | 9 | 8852 | ||
| XM_017013883.1 | XP_016869372.1 | 9 | 8952 | |||
| XM_017013885.1 | XP_016869374.1 | 9 | 8660 | |||
| GenBank mRNA | AK022971.1 | - | 1 | 3208 | ||

SLCO5A1
Genomic Map