siRNA Details
| Gene Symbol: LOC401433 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK127717.1 | - | 1 | 777 | ||
| Gene Symbol: WDR86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011516144.2 | XP_011514446.1 | 7 | 2484 | ||
| XM_011516145.2 | XP_011514447.1 | 6 | 2355 | |||
| XM_011516147.2 | XP_011514449.1 | 6 | 2284 | |||
| XM_011516151.2 | XP_011514453.1 | 5 | 2155 | |||

LOC401433
Genomic Map