siRNA Details
Gene Symbol: ZNF549 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL833090.2 | - | 4 | 4833 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK128156.1 | - | 1 | 2165 | Chr. 9 - 137114250 - 137117138 [+] on Build GRCh38 |
