siRNA Details
Gene Symbol: LINC00536 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046215.1 | 5 | 1363 | |||
GenBank mRNA | BC040336.1 | - | 5 | 1363 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00536 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046215.1 | 5 | 1363 | |||
GenBank mRNA | BC040336.1 | - | 5 | 1363 |