siRNA Details
| Gene Symbol: SFMBT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001018039.1 | NP_001018049.1 | 21 | 6270 | ||
| NM_001029880.2 | NP_001025051.1 | 21 | 6372 | |||
| XM_005252551.2 | XP_005252608.1 | 21 | 6153 | |||
| XM_006717490.2 | XP_006717553.1 | 19 | 6145 | |||
| XM_011519611.1 | XP_011517913.1 | 21 | 6371 | |||
| XM_017016466.1 | XP_016871955.1 | 21 | 6273 | |||
| GenBank mRNA | AK057130.1 | - | 1 | 1490 | ||

SFMBT2
Genomic Map