siRNA Details
| Gene Symbol: EDN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC036851.1 | - | 1, 2 | 358 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EDN1| Gene Symbol: EDN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC036851.1 | - | 1, 2 | 358 | ||