siRNA Details
| Gene Symbol: SPTBN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005274192.4 | XP_005274249.1 | 38 | 9577 | ||
| XM_005274193.3 | XP_005274250.1 | 37 | 9351 | |||
| XM_006718669.3 | XP_006718732.1 | 36 | 11264 | |||
| XM_006718671.3 | XP_006718734.1 | 38 | 8977 | |||
| XM_017018174.1 | XP_016873663.1 | 38 | 9669 | |||
| XM_017018175.1 | XP_016873664.1 | 38 | 9298 | |||
| XM_017018176.1 | XP_016873665.1 | 38 | 9018 | |||
| XM_017018177.1 | XP_016873666.1 | 37 | 9207 | |||
| XM_017018178.1 | XP_016873667.1 | 37 | 8890 | |||
| GenBank mRNA | AK021479.1 | - | 1 | 1063 | ||

SPTBN2
Genomic Map