siRNA Details
| Gene Symbol: SNX24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021473.1 | - | 1 | 736 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CB111567.1 | - | 1 | 280 | Chr. 5 - 123007059 - 123007618 [+] on Build GRCh38 | |

SNX24
Genomic Map