siRNA Details
| Gene Symbol: LOC105373495 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC015425.1 | - | 1 | 1054 | ||
| BC053735.1 | - | 1 | 1079 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105373495| Gene Symbol: LOC105373495 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC015425.1 | - | 1 | 1054 | ||
| BC053735.1 | - | 1 | 1079 | |||