siRNA Details
| Gene Symbol: FLJ13773 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK023835.1 | - | 1 | 2295 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FLJ13773| Gene Symbol: FLJ13773 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK023835.1 | - | 1 | 2295 | ||