siRNA Details
| Gene Symbol: RNF165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK091537.1 | - | 1 | 1624 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNF165| Gene Symbol: RNF165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK091537.1 | - | 1 | 1624 | ||