siRNA Details
| Gene Symbol: KLF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001324105.1 | NP_001311034.1 | 1 | 144 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CD387915.1 | - | 65 | Chr. - - [-] on Build | ||

KLF8
Genomic Map