siRNA Details
| Gene Symbol: ZMIZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK097263.1 | - | 1 | 1434 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX118977.1 | - | 1 | 124 | Chr. 7 - 44771170 - 44771767 [+] on Build GRCh38 | |

ZMIZ2
Genomic Map