siRNA Details
| Gene Symbol: FRMD4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK093978.1 | - | 1 | 1645 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | EL955225.1 | - | 1 | 210 | Chr. 10 - 13689966 - 13690352 [-] on Build GRCh38 | |

FRMD4A
Genomic Map