siRNA Details
| Gene Symbol: LOC283140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126004.1 | 4 | 577 | |||
| GenBank mRNA | AK095275.1 | - | 4 | 581 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA440806.1 | - | 4 | 581 | Chr. 11 - 112270749 - 112360397 [+] on Build GRCh38 | |

LOC283140
Genomic Map