siRNA Details
| Gene Symbol: APOPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001302654.1 | NP_001289583.1 | 4 | 613 | ||
| GenBank mRNA | BX119699.1 | - | 4 | 493 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK024310.1 | - | 1 | 259 | Chr. 14 - 103574317 - 103714982 [+] on Build GRCh38 | |

APOPT1
Genomic Map