siRNA Details
| Gene Symbol: COL4A3BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011543090.2 | XP_011541392.1 | 17 | 5726 | ||
| GenBank mRNA | AK125387.1 | - | 1 | 1205 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

COL4A3BP| Gene Symbol: COL4A3BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011543090.2 | XP_011541392.1 | 17 | 5726 | ||
| GenBank mRNA | AK125387.1 | - | 1 | 1205 | ||