siRNA Details
| Gene Symbol: PAWR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006719436.3 | XP_006719499.1 | 5 | 3415 | ||
| GenBank mRNA | AK027848.2 | - | 1 | 916 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PAWR| Gene Symbol: PAWR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006719436.3 | XP_006719499.1 | 5 | 3415 | ||
| GenBank mRNA | AK027848.2 | - | 1 | 916 | ||